MadgeTech RHTEMP1000 Temp/RH Recorder
Representative image only.
MadgeTech RHTEMP1000 Temp/RH Recorder
MadgeTech | Mfr: RHTEMP1000| Item: 1800342
Call For Price
for an additional
₹40,300.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
MadgeTech RHTEMP1000 Temp/RH Recorder
MadgeTech RHTEMP1000 Temp/RH Recorder
1800342
Call For Price
+ =
Total Price 40300.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Real-time operation
  • Low cost
  • User-friendly
  • Battery powered - user replaceable
  • Programmable start time
  • Reusable
  • Small size
  • Non-volatile solid state memory
  • Readings selectable 1 every 2 sec to 1 every 12 hours
  • 1 year warranty
More About this Item
The MH1000 series data loggers can be started and stopped directly from a personal computer and its small size allows it to fit almost anywhere. Data retrieval is quick and easy. Simply plug a logger into an empty COM port and our user-friendly software allows data to be examined, exported and printed.
Conformite Europeenne1 Year Warranty
Specifications
  • Number of Channels1
  • Input TypeTemperature and humidity
  • Max Temperature (° F)176
  • Min Temperature (° F)-40
  • Min Temperature (° C)-40
  • Max Temperature (° C)80
  • DescriptionSubmersible Temp/RH Recorder