M-System MDP-24T Lightning/Surge Protector; 4-20mA, Pulse, Photovoltaic
Representative image only.
M-System MDP-24T Lightning/Surge Protector; 4-20mA, Pulse, Photovoltaic
M-System | Mfr: MDP-24T| Item: 4300202
Don’t let lightning take systems down
₹25,800.00INR / Each
₹25,800.00INR / Each
Product Description
Key Features
  • Absorb only the lightning surges with no interruption of the instrumentation signal
  • Lightweight, easy-to-handle, plug-in construction
  • Superior protection and selection across a wide range of applications
  • Remove and test without disconnecting wires
  • Base socket connects input-output signals when an element is removed
  • 3 year warranty
3 Year Warranty
Specifications
  • DescriptionLightning/Surge Protector; 4-20mA, Pulse, Photovoltaic
  • Warranty3 Years