M-System M2SED-A13-R/UL Dc Input Limit Alarm Thin Profile 4-20 Ma Dual Spdt
Representative image only.
M-System M2SED-A13-R/UL Dc Input Limit Alarm Thin Profile 4-20 Ma Dual Spdt
M-System | Mfr: M2SED-A13-R/UL| Item: 6846764
Monitor a range of alarm conditions
₹66,000.00INR / Each
₹66,000.00INR / Each
Product Description
Key Features
  • Thumbwheel switch adjustments
  • Only 1.2 inch (29.5mm) wide
  • Front LCD display indicating values in scaled engineering unit range is used to program the module
  • Quad or dual trip
  • Program Lockout function
  • Front-panel configurable
  • 3 year warranty
More About this Item
–– Thumbwheel switch adjustments
–– Only 1.2 inch (29.5mm) wide
3 Year Warranty
Specifications
  • Relay TypeSPDT
  • DescriptionDc Input Limit Alarm Thin Profile 4-20 Ma Dual Spdt
  • Warranty3 Years