Lovibond TB350 T-Cal® Standard Set B, For TB350 IR
Representative image only.
Lovibond TB350 T-Cal® Standard Set B, For TB350 IR
Lovibond | Mfr: 194154| Item: 9959988
Calibrate, clean, and keep your TB350 turbidity meters running smoothly
₹86,400.00INR / Each
₹86,400.00INR / Each
Product Description
Includes
5, 20, 800, 4000 NTU standards.
Key Features
  • Stabilized Formazin turbidity standard in ready-to-use sealed vials
  • Save time and eliminate errors - no need for dilution
  • US EPA and ISO approved as primary calibration standards
More About this Item
This set delivers all of the concentrations needed to calibrate and verify the TB 350 IR Portable Turbidimeter. The standards are supplied in sealed vials, eliminating any need for preparation.
T-CAL® Turbidity Standards are stable Formazin standards prepared in ready-to-use concentrations to ensure accurate turbidity measurements. All concentrations are verified to meet tight quality control specifications, are EPA & ISO compliant for reporting purposes and are guaranteed to save you time.
Specifications
  • DescriptionT-Cal® Standard Set B, For TB350 IR