Lovibond TB350 T-Cal® Standard Set A, For TB350 WL
Representative image only.
Lovibond TB350 T-Cal® Standard Set A, For TB350 WL
Lovibond | Mfr: 194152| Item: 9959987
Calibrate, clean, and keep your TB350 turbidity meters running smoothly
₹86,400.00INR / Each
₹86,400.00INR / Each
Product Description
Includes
5, 20, 800, 2000, 4000 NTU standards.
Key Features
  • Stabilized Formazin turbidity standard in ready-to-use sealed vials
  • Save time and eliminate errors - no need for dilution
  • US EPA approved as primary calibration standards
More About this Item
This set delivers all of the concentrations needed to calibrate and verify the TB 350 WL Portable Turbidimeter. The standards are supplied in sealed vials, eliminating any need for preparation and ensuring safe operation.
T-CAL® Turbidity Standards are stabilized Formazin standards prepared in ready-to-use concentrations to ensure accurate turbidity measurements. All concentrations are verified to meet tight quality control specifications, are EPA compliant for reporting purposes and are guaranteed to save time.
Specifications
  • DescriptionT-Cal® Standard Set A, For TB350 WL