Lovibond TB350 Silicone Oil
Representative image only.
Lovibond TB350 Silicone Oil
Lovibond | Mfr: 194295| Item: 9959997
Calibrate, clean, and keep your TB350 turbidity meters running smoothly
₹4,100.00INR / Each
₹4,100.00INR / Each
Product Description
More About this Item
Silicone oil can be used to prepare sample cells for turbidity measurement. The oil can be used to mask small scratches and other imperfections in the glassware.
Specifications
  • DescriptionSilicone Oil