Lovibond ST100-10 Primary Chlorine Standard Kit for Colorimeters
Representative image only.
Lovibond ST100-10 Primary Chlorine Standard Kit for Colorimeters
Lovibond | Mfr: 48105510| Item: 9956090
Validate the calibration and accuracy of your instrument
₹11,000.00INR / Each
₹11,000.00INR / Each
Product Description
Includes
Chlorine standard ampoule, 97 mL of zero chlorine water, and ampoule breaker.
Key Features
  • Complies with new regulations specified in US EPA Method 334.0
  • Ideal for laboratory and on-line chlorine measurements
  • 1 year warranty
More About this Item
Everything you need for easy preparation of a 1.5 mg/L NIST traceable chlorine standard. Simple, time saving and practical kit to help comply with the new regulations.
1 Year Warranty
Specifications
  • DescriptionPrimary Chlorine Standard Kit for Colorimeters