Lovibond RP012-3 Powder Pillow, DPD Total Chlorine Reagent, 10 mL, 1000 tests
Representative image only.
Lovibond RP012-3 Powder Pillow, DPD Total Chlorine Reagent, 10 mL, 1000 tests
Lovibond | Mfr: 530123| Item: 9956059
Premeasured reagents in convenient powder packs
₹90,600.00INR / Pkg of 1
₹90,600.00INR / Pkg of 1
Product Description
Key Features
  • Precise dosing with simple push-button operation
More About this Item
These powder packs contain premeasured unit dose reagents suitable for use with Lovibond/Orbeco-Hellige colorimeters, Aqua comparator test kits, YSI, Thermo, and Hach® instrumentation/test kits. Reagents for chlorine are DPD chemistry. Some parameters require multiple reagents and are bundled into a "Reagent set", which can include powder packs, tube tests, or both.
Specifications
  • Test ParametersDPD total chlorine
  • Number of Tests Included1000 x 10 mL
  • DescriptionPowder Pillow, DPD Total Chlorine Reagent, 10 mL, 1000 tests