Lovibond RP009-0 Powder Pillow, DPD Free Chlorine Reagent, 5 mL, 100 tests
Representative image only.
Lovibond RP009-0 Powder Pillow, DPD Free Chlorine Reagent, 5 mL, 100 tests
Lovibond | Mfr: 530090| Item: 9956052
Premeasured reagents in convenient powder packs
₹9,900.00INR / Pkg of 1
₹9,900.00INR / Pkg of 1
Product Description
Key Features
  • Precise dosing with simple push-button operation
More About this Item
These powder packs contain premeasured unit dose reagents suitable for use with Lovibond/Orbeco-Hellige colorimeters, Aqua comparator test kits, YSI, Thermo, and Hach® instrumentation/test kits. Reagents for chlorine are DPD chemistry. Some parameters require multiple reagents and are bundled into a "Reagent set", which can include powder packs, tube tests, or both.
Specifications
  • Test ParametersDPD free chlorine
  • Number of Tests Included100 x 5 mL
  • DescriptionPowder Pillow, DPD Free Chlorine Reagent, 5 mL, 100 tests