Lovibond PFXi-195/1 Short Pathlength Color Meter, Liquid Chemicals and Oils
Representative image only.
Lovibond PFXi-195/1 Short Pathlength Color Meter, Liquid Chemicals and Oils
Lovibond | Mfr: 1371951| Item: 9956202
Eliminate user subjectivity with unbiased color measurements based on reference standards
Call For Price
Call For Price
Buy It Together
Current Item
Lovibond PFXi-195/1 Short Pathlength Color Meter, Liquid Chemicals and Oils
Lovibond PFXi-195/1 Short Pathlength Color Meter, Liquid Chemicals and Oils
9956202
Call For Price
+
Lovibond 606200 Replacement Optical Glass Cell, 50 mm
9956255
₹58,800.00INR / Each
=
Total Price 58800.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
optical glass cell, data analysis software, calibration filter(s) with certificate, spare lamp, and 10 ft (3 m) power cable with US, UK, and EU plugs.
Key Features
  • Large graphic LCD with user-friendly prompts in several languages
  • Extensive choice of standard color scales or customized color scales from reference samples
  • Easily removable sample chamber offering a range of sample cell sizes
  • Data output meets GLP standards with date, time, sample, and user ID
  • CIE L*a*b
  • Gardner Color
  • Iodine Color (DIN 6162)
  • Platinum-Cobalt/Hazen/APHA Color (ASTM D 1209)
  • xyY chromaticity co-ordinates
  • XYZ tristimulus values
  • Optical density (Absorbance)
  • A Guide to Path Length by Colour Scale
  • Delta E* color difference
  • Spectral data
  • 1 year warranty
More About this Item
Spectrophotometric color meters provide accurate, reliable color data from optically clear samples. The meter automatically measures color transmittance and displays results directly in industry color scales, CIE values, and as spectral data. The direct measurements on large LCD supply unbaised readings, unaffected by operator or environment.
The short-pathlength meter is ideal for slightly colored solutions, like wastewater, syrups, honey, dark oils, and beers. The meter offers built-in menu guides to help navigate setup parameters for the different color scales. The meter performance is verified with self-diagnostic check tests, and accuracy is ensured with calibrations using certified glass filter(s) included with meter (filters differ depending upon model).
Flexible data transfer with USB, LAN, and RS-232 interfaces. Remote calibration and maintenance service are also available via the internet. Contact our Applications Specialists for upgrades to color scales.
1 Year Warranty
Specifications
  • TestLiquid chemicals and industrial oils
  • DescriptionShort Pathlength Color Meter, Liquid Chemicals and Oils