Lovibond MD100 Colorimeter Manganese LR Kit

Representative image only.

Lovibond MD100 Colorimeter Manganese LR Kit

Lovibond | Mfr: 276105| Item: 9956188

Uses popular Powder Pack™, tablet, and liquid chemistries

Call For Price

Call For Price
Product Description

Includes

three 24-mm sample cells with light-shield caps, reagents for 100 tests, foam-lined case, and one 9V battery.

Key Features

  • Large backlit display for low light conditions
  • Sixteen-point automatic data storage with time and date stamp
  • Ergonomic, waterproof design for easy hand-held use in lab, field, or plant
  • 2 year warranty

More About this Item

The Lovibond MD100 series colorimeters are available in a wide variety of parameters using powder-pack chemistries. These rugged, waterproof meters feature a large backlit display, real-time clock, data logging capabilities, and a user calibration mode which allows you to calibrate against prepared standards.

(Note: Lovibond is the new name for Orbeco-Hellige--exactly the same product with a new name.)

Range for reagent: 0.011 to 0.7 mg/L

Two year warrantyConformite Europeenne
Specifications
  • Display TypeLCD
  • Light SourceLED
  • Absorbance Accuracy±1 nm
  • Test ParametersManganese; low-range
  • Min Operating Temperature (° F)32
  • Max Operating Temperature (° F)104
  • Min Operating Temperature (° C)0
  • Max Operating Temperature (° C)40
  • Number of Tests Included100
  • Data LoggingYes
  • Memory Size16 data sets
  • Battery1 9V (included)
  • Length (in)6 1/8
  • Width (in)3
  • Height (in)1 1/2
  • Range0.01 to 0.70 mg/L
  • DescriptionColorimeter Manganese LR Kit
  • Warranty2 year
Reviews
Q&A