Lovibond MD100 Colorimeter, Chlorine/pH Kit

Representative image only.

Lovibond MD100 Colorimeter, Chlorine/pH Kit

Lovibond | Mfr: 278030| Item: 9956152

Uses popular Powder Pack™, tablet, and liquid chemistries

₹167,000.00INR / Each

₹167,000.00INR / Each

Buy It Together

Current Item

Lovibond MD100 Colorimeter, Chlorine/PH Kit
Lovibond MD100 Colorimeter, Chlorine/PH Kit
9956152

₹167,000.00INR / Each
+ =
Total Price 177500.00 ADD 2 ITEMS TO CART
Product Description

Includes

three 24-mm sample cells with light-shield caps, reagents for 100 tests, foam-lined case, and one 9V battery.

Key Features

  • Large backlit display for low light conditions
  • Sixteen-point automatic data storage with time and date stamp
  • Ergonomic, waterproof design for easy hand-held use in lab, field, or plant
  • 2 year warranty

More About this Item

The Lovibond MD100 series colorimeters are available in a wide variety of parameters using powder-pack chemistries. These rugged, waterproof meters feature a large backlit display, real-time clock, data logging capabilities, and a user calibration mode which allows you to calibrate against prepared standards.

(Note: Lovibond is the new name for Orbeco-Hellige--exactly the same product with a new name.)

Two year warranty
Specifications
  • Display TypeLCD
  • Light SourceLED
  • Test ParameterspH and chlorine
  • Min Operating Temperature (° F)32
  • Max Operating Temperature (° F)104
  • Min Operating Temperature (° C)0
  • Max Operating Temperature (° C)40
  • Number of Tests Included100
  • WaterproofYes
  • Data LoggingYes
  • Battery1 9V (included)
  • Length (in)6 3/4
  • Width (in)2 5/8
  • Height (in)1 13/16
  • Length (cm)17
  • Width (cm)6.5
  • Height (cm)4.5
  • Range0.01 to2.0 mg/L, 6.5 to 8.4 pH
  • DescriptionColorimeter, Chlorine/pH Kit
  • Warranty2 year
Reviews
Q&A