Lovibond Calibration Set for TB200 Turbidity Meters
Representative image only.
Lovibond Calibration Set for TB200 Turbidity Meters
Lovibond | Mfr: 194280| Item: 9959996
Optimize water analysis
₹105,000.00INR / Each
₹105,000.00INR / Each
Product Description
Key Features
  • For Lovibond water analysis equipment
More About this Item
Choose the following water analysis accessories to meet your application needs.
Contains one sealed vial each of 0.02, 10, and 1000 NTU.
Specifications
  • Compatible WithOrbeco-Hellige TB200 turbidity meters
  • DescriptionCalibration Set for TB200 Turbidity Meters