Lighting Specialties Replacement Bulb for Infrared Heat Lamp, 250 watts
Representative image only.
Lighting Specialties Replacement Bulb for Infrared Heat Lamp, 250 watts
Lighting Specialties | Mfr: 250R40/HR/1| Item: 0305701
Fast, safe heat without airflow
₹15,500.00INR / Each
₹15,500.00INR / Each
Product Description
Key Features
  • Durable all-metal construction
  • Adjustable 42" swivel extension arm
  • Inter-reflector prevents excessive heat build-up
  • 1 year warranty
More About this Item
Mount to any table or benchtop, or use our floor stands to support the heating lamp—features durable all-metal construction. Adjust the 42" swiveling extension arm to position lamp exactly where you want it. Dual reflector prevents excessive heat buildup.
Lamp comes complete with screw-type mounting bracket; 10-ft grounded cord with plug; and 250 watt, hard glass infrared heat bulb. Easily remove wire bulb guard to replace bulb (order replacement bulb separately). Various base stands are available (order separately).
1 Year Warranty
Specifications
  • DescriptionReplacement bulb for infrared heating lamp, 250 watts