Lascar Electronics EL-USB-2-LCD Data Logger, RH/Temp/Dew Pt, with LCD Display
Representative image only.
Lascar Electronics EL-USB-2-LCD Data Logger, RH/Temp/Dew Pt, with LCD Display
Lascar Electronics | Mfr: EL-USB-2-LCD| Item: 2303962
Direct-to-USB connection and easy-to-use functionality
Call For Price
for an additional
₹40,300.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
Lascar Electronics EL-USB-2-LCD Data Logger, RH/Temp/Dew Pt, with LCD Display
Lascar Electronics EL-USB-2-LCD Data Logger, RH/Temp/Dew Pt, with LCD Display
2303962
Call For Price
+ + =
Total Price 40300.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • User-programmable alarm thresholds
  • All data loggers come with software and are great for laboratory environmental monitoring
  • 1 year warranty
More About this Item
Simply insert any of these compact, 4.5" (11.4 cm) long loggers into your computer USB port, name it, choose your required sampling rate, select high and low alarms if required, and set the logger start time. Once configured, the logger can be removed from the computer to log independently. Data is downloaded back onto the PC where it can be analyzed and viewed as a graph using included software.
These data loggers measure temperature and humidity from 31 to 176°F (–35 to 80°C) and 0 to 100% RH, calculate dew point, and store over 16,000 readings per parameter.
Model EL-USB-2-LCD features a 2.5-digit LCD showing the latest logged values, min/max log, and have immediate, delayed, and push-to-start logging.
What's included: configuration software and battery.
1 Year Warranty
Specifications
  • Number of Channels1
  • Input TypeTemperature, dew point, and humidity
  • Max Temperature (° F)158
  • Min Temperature (° F)-4
  • Max Temperature (° C)70
  • Min Temperature (° C)-20
  • Temperature Resolution (° F)1
  • Temperature Accuracy (° F)±2.5F
  • Temperature Resolution (° C)0.5
  • Max Humidity RH (RH)100
  • Min Humidity RH (RH)0
  • Humidity Resolution (%)0.5
  • Humidity Accuracy±3% RH
  • Display TypeLCD
  • DescriptionData Logger, RH/Temp/Dew Pt, with LCD Display