LaMotte 7191-02 Liquid Sanitizer Strength Verification Kits, Peracetic Acid Test
Representative image only.
LaMotte 7191-02 Liquid Sanitizer Strength Verification Kits, Peracetic Acid Test
LaMotte | Mfr: 7191-02| Item: 9953206
Titrimetric kits verify the concentration of your sanitizer quickly and easily
Call For Price
Call For Price
Product Description
Key Features
  • Save money by avoiding overuse or underuse of sanitizers
  • 1 year warranty
More About this Item
Verify the strength and effectiveness of your liquid sanitizer with these colorimetric test kits. All kits contain reagents for 50 tests.
1 Year Warranty
Specifications
  • TestPeracetic acid
  • Measurement Points50 ppm/drop peroxide; 15 ppm/drop peracetic acid
  • Tests Per Kit50
  • DescriptionLiquid Sanitizer Strength Verification Kits, Peracetic Acid Test