LaMotte 6973-H Colorimeter Chlorine Standard, 250 ppm; 60 Ml

Representative image only.

LaMotte 6973-H Colorimeter Chlorine Standard, 250 ppm; 60 Ml

LaMotte | Mfr: 6973-H| Item: 0556416

Optimize performance of your LaMotte® colorimeter and turbidity meters

Call For Price

Call For Price

Buy It Together

Current Item

LaMotte 6973-H Colorimeter Chlorine Standard, 250 ppm; 60 Ml
LaMotte 6973-H Colorimeter Chlorine Standard, 250 ppm; 60 Ml
0556416

Call For Price
+ =
Total Price 3500.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Replacement parts, accessories, and reagents to keep your colorimeter and turbidity results dependable

More About this Item

Choose from a wide range of accessories and reagents to measure water testing parameters utilizing LaMotte® colorimeter and turbidity meters. Meters sold separately.

Specifications
  • Test ParametersChlorine
  • Range250 ppm
  • DescriptionColorimeter Chlorine Standard, 250 ppm; 60 mL
Documents & Certificates
Reviews
Q&A