LaMotte 5010-01 Soil Nutrient Test Kit for Macronutrient, Micronutrient, and pH; 50 Tests
Representative image only.
LaMotte 5010-01 Soil Nutrient Test Kit for Macronutrient, Micronutrient, and pH; 50 Tests
LaMotte | Mfr: 5010-01| Item: 9903110
Quick, visible color-change indicates results
Call For Price
Call For Price
Buy It Together
Current Item
LaMotte 5010-01 Soil Nutrient Test Kit for Macronutrient, Micronutrient, and pH; 50 Tests
LaMotte 5010-01 Soil Nutrient Test Kit for Macronutrient, Micronutrient, and pH; 50 Tests
9903110
Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART
Product Description
Includes
test reagents, a soil management handbook for determining lime and fertilizer requirements, and a pad of soil analysis report forms.
Key Features
  • Refillable reagents minimize product usage costs
  • 1 year warranty
More About this Item
Soil test kits simplify measuring the pH level, nutrients, or texture of your soil. This line of combination soil analysis outfits offers the finest visual color matching system available to today’s agronomist. Technically advanced reagent systems and unique extraction procedures based on the Mehlich I extraction provide fast, simple and extremely accurate soil testing. A series of rapid, accurate chemical tests use standardized reagents to produce color reactions measured against laminated color charts.
Tests for calcium, chloride, potassium, and sulfate are based on turbidity measurements. Refill reagents are available to replenish your kit.
Soil test kits simplify measuring the pH level, macronutrient, and micronutrient levels of your soil. Tests use standard reagents to produce color changes; compare results to color charts for concentration determination. Tests for Aluminum, Calcium, Chloride, Humus (organic), Iron (ferric), Magnesium, Manganese, Nitrogen (ammonia), Nitrogen (nitrate), Nitrogen (nitrite), pH, Phosphorus, Potassium, and Sulfate.
1 Year Warranty
Specifications
  • Test Range Aluminum5-125 ppm
  • Test Range Calcium150-2800 ppm
  • Test Range Chloride25-500 ppm
  • Test Range Humus (organic)1.5-8%
  • Test Range Iron (ferric)5-125 lbs/acre
  • Test Range Magnesium5-150 ppm
  • Test Range Manganese4-40 ppm
  • Test Range Nitrogen (ammonia)5-150 ppm
  • Test Range Nitrogen (nitrate)10-150 lbs/acre
  • Test Range Nitrogen (nitrite)1-50 ppm
  • Test Range pH3.8-9.6 pH
  • Test Range Phosphorus10-200 lbs/acre
  • Test Range Potassium100-400 lbs/acre
  • Test Range Sulfate50-2000 ppm
  • Number of Tests Aluminum50
  • Number of Tests Calcium50
  • Number of Tests Chloride50
  • Number of Tests Humus (organic)50
  • Number of Tests Iron (ferric)50
  • Number of Tests Magnesium50
  • Number of Tests Manganese50
  • Number of Tests Nitrogen (ammonia)50
  • Number of Tests Nitrogen (nitrate)50
  • Number of Tests pH100
  • Number of Tests Phosphorus50
  • Number of Tests Potassium50
  • Number of Tests Sulfate50
  • DescriptionSoil Nutrient Test Kit for Macronutrient, Micronutrient, and pH; 50 Tests