LaMotte 4140-03 DPD Chlorine Secondary Standards Kit
Representative image only.
LaMotte 4140-03 DPD Chlorine Secondary Standards Kit
LaMotte | Mfr: 4140-03| Item: 0556412
Optimize performance of your LaMotte® colorimeter and turbidity meters
₹43,100.00INR / Each
₹43,100.00INR / Each
Buy It Together
Current Item
LaMotte 4140-03 DPD Chlorine Secondary Standards Kit
LaMotte 4140-03 DPD Chlorine Secondary Standards Kit
0556412
₹43,100.00INR / Each
+ =
Total Price 46600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Replacement parts, accessories, and reagents to keep your colorimeter and turbidity results dependable
More About this Item
Choose from a wide range of accessories and reagents to measure water testing parameters utilizing LaMotte® colorimeter and turbidity meters. Meters sold separately.
Specifications
  • Test ParametersChlorine
  • RangeStandard A: 0.1 to 0.4 ppm; Standard B: 0.9 to 1.2 ppm; Standard C 2.2 to 3.2 ppm
  • DescriptionDPD Chlorine Secondary Standards Kit