LaMotte 4-3616 Test Kit , Coliform, 5 Tests/Kit
Representative image only.
LaMotte 4-3616 Test Kit , Coliform, 5 Tests/Kit
LaMotte | Mfr: 4-3616| Item: 9956100
Test kit screens for coliform bacterial contamination
₹13,000.00INR / Each
₹13,000.00INR / Each
Product Description
Key Features
  • Store at room temperature—no incubator needed
More About this Item
Simply pour sample into bag, shake to dissolve dechlorinating tablet, pour into the five glass tubes, and store tubes at room temperature. Color change and small bubble formation provide indication of bacterial presence.
What's included: sterile water sampling bag with dechlorinating tablet, five 10-mL glass tubes, and five coliform test tablets.
Specifications
  • TestBacteria, coliform
  • Tests Per Kit5
  • DescriptionTest Kit , Coliform, 5 Tests/Kit