LaMotte 1067 Soil Texture Test Kit for Sand, Silt, and Clay Fractions; 50 Tests
Representative image only.
LaMotte 1067 Soil Texture Test Kit for Sand, Silt, and Clay Fractions; 50 Tests
LaMotte | Mfr: 1067| Item: 9903000
Quick, visible color-change indicates results
₹32,300.00INR / Each
₹32,300.00INR / Each
Product Description
Includes
soil flocculating reagent 60 mL, texture dispersing reagent 60 mL, soil texture stand, three 50 mL test tubes with caps, two 1 mL plastic pipettes with caps, and a soil management handbook for determining lime and fertilizer requirements.
Key Features
  • Refillable reagents minimize product usage costs
  • 1 year warranty
More About this Item
Soil test kits simplify measuring the pH level, nutrients, or texture of your soil. This line of combination soil analysis outfits offers the finest visual color matching system available to today’s agronomist. Technically advanced reagent systems and unique extraction procedures based on the Mehlich I extraction provide fast, simple and extremely accurate soil testing. A series of rapid, accurate chemical tests use standardized reagents to produce color reactions measured against laminated color charts.
Tests for calcium, chloride, potassium, and sulfate are based on turbidity measurements. Refill reagents are available to replenish your kit.
The overall texture of a soil affects growth in the root zone, which determines the above-ground growth production, and is determined by the fractions of sand, silt, and clay present. Test determines sand, silt, and clay fraction soil texture.
1 Year Warranty
Specifications
  • DescriptionSoil Texture Test Kit for Sand, Silt, and Clay Fractions; 50 Tests