LaMotte 0171-CL Secchi Turbidity Disk, 20 m cable
Representative image only.
LaMotte 0171-CL Secchi Turbidity Disk, 20 m cable
LaMotte | Mfr: 0171-CL| Item: 0549200
Use to determine turbidity or degree of visibility in natural water supplies
₹35,400.00INR / Each
₹35,400.00INR / Each
Product Description
Key Features
  • Braided, stretch resistant line marked at half and full meter intervals up to 20 meters
  • 1 year warranty
More About this Item
Simple-to-use secchi disk is a weighted, 20-cm dia black and white plate that sinks parallel to the water's surface. For best results use, from the leeward side of a boat or dock between mid-morning and mid-afternoon.
1 Year Warranty
Specifications
  • Meter TypePortable
  • DescriptionSecchi Turbidity Disk, 20 m cable