Labmat F246751000 20" X 50'FT roll, Orange Biohazard Designated Liner

Representative image only.

Labmat F246751000 20" X 50'FT roll, Orange Biohazard Designated Liner

Labmat | Mfr: F246751000| Item: 0680503

Skid resistent polyethylene film backing that prevents leakage

₹32,400.00INR / Roll of 1

₹32,400.00INR / Roll of 1
Product Description

Key Features

  • Disposable

More About this Item

Highly abosrbent top layer soaks up corrosive materials, and most liquids. Red backing alerts end-users when biohazard materials are in use. Liners can easily be cut to meet end users specific applications.

Specifications
  • MaterialPolyethylene
  • Width (in)20
  • Length (ft)50
  • Width (cm)50.8
  • Length (m)15.24
  • ColorOrange
  • Description20" X 50'FT roll, Orange Biohazard Designated Liner
Reviews
Q&A