Labconco Silencer for Scroll Vacuum Pumps
Representative image only.
Labconco Silencer for Scroll Vacuum Pumps
Labconco | Mfr: 7587300| Item: 8888890
Reduces exhaust pump noise
₹154,000.00INR / Each
₹154,000.00INR / Each
Product Description
Includes
Filter, centering ring, and clamp.
Key Features
  • Ideal for use with Scroll vacuum pumps
  • 1 year warranty
More About this Item
Use this silencer to filters exhaust while silencing exhaust pump noise from Labconco Scroll vacuum pumps. It also prevents tip seal dust from contaminating the environment. The sound absorbent element is replaceable. Includes filter, centering ring and clamp to attach to exhaust outlet.
1 Year Warranty
Specifications
  • DescriptionLabconco 7587300 Silencer for Scroll Vac Pump