Labconco Purifier® HEPA Filter for 72" Purifier® Horizontal Clean Benches
Representative image only.
Labconco Purifier® HEPA Filter for 72" Purifier® Horizontal Clean Benches
Labconco | Mfr: 1489004| Item: 0911686
Ensure clean airflow with filters and prefilters
₹728,000.00INR / Each
₹728,000.00INR / Each
Product Description
Key Features
  • Fits in clean benches to help protect critical, non-hazardous samples
  • Compatible with 72" Purifier® horizontal clean benches
  • 1 year warranty
Specifications
  • Compatible With72" Purifier® horizontal clean benches
  • Width (InchShort)72
  • TypeHEPA filter
  • DescriptionHEPA Filter for 72" Purifier® Horizontal Clean Benches