Labconco Protector Downdraft Powder Station, 3-ft Wide, 115 V

Representative image only.

Labconco Protector Downdraft Powder Station, 3-ft Wide, 115 V

Labconco | Mfr: 3410001| Item: 2861606

Fingerprint powdering of large items is no longer a challenge!

Call For Price

Call For Price
Product Description

Includes

prefilter, HEPA filter, stainless steel worksurface, and 10-ft (3-m) power cord

Key Features

  • Safely work with nuisance powders
  • Open-side, open-top design allows unrestricted work space and accommodates oversized items
  • 1 year warranty

More About this Item

The HEPA filtered, open design of the downdraft powder stations safely contain powders while giving the freedom of an open work area. With the addition of the optional transition connector 28616-38, multiple downdraft powder stations can be joined together to create a longer, continuous work station.

Durable workstations are constructed of epoxy coated steel and include a removable stainless steel work surface. Beneath the work surface is a powder collection tray that is removable for easy clean up. The 75-watt blower is adjustable to optimize the life of each HEPA filter. A filter alarm light alerts you to low airflow conditions prompting a filter change. Each downdraft powder station comes complete with a prefilter and a HEPA filter; available in 2-ft (0.6-m) and 3-ft (0.9-m) widths

Note This product is for nuisance powders only. It is not to be used for biohazards, toxic chemicals or radioisotopes.

Conformite Europeenne1 Year Warranty
Specifications
  • Hood Width (in)36
  • Exterior Width (in)36
  • Exterior Height (in)13.3125
  • Exterior Depth (in)25.8125
  • Power (VAC)115
  • Power (Hz)60
  • Filter TypeHEPA
  • DescriptionDowndraft Powder Station, 3-ft Wide, 115 V
  • Warranty1 Year
Reviews
Q&A