Labconco Logic Vue Safety Interlock Cable
Representative image only.
Labconco Logic Vue Safety Interlock Cable
Labconco | Mfr: 3349700| Item: 3351785
Enhance the performance of your enclosure with the following accessories
₹7,500.00INR / Each
₹7,500.00INR / Each
Product Description
Key Features
  • Connect the enclosure and the automated devices that employ a safety interlock
  • 1 year warranty
More About this Item
A safety interlock pauses automated instruments when operating conditions are unsafe. Opening the sash of the Logic® Vue enclosure triggers the interlock, immediately signaling a device to pause or stop.
1 Year Warranty
Specifications
  • Compatible WithAll Logic® Vue biosafety enclosures
  • DescriptionSafety Interlock Cable