Labconco Logic Vue Large Tubing Passthrough Port, 1-1/4"
Representative image only.
Labconco Logic Vue Large Tubing Passthrough Port, 1-1/4"
Labconco | Mfr: 3349201| Item: 3351784
Enhance the performance of your enclosure with the following accessories
₹59,300.00INR / Each
₹59,300.00INR / Each
Product Description
Key Features
  • Allows passage of tubing up to 1-1/2" through sidewall of Logic® Vue enclosure for external equipment
  • 1 year warranty
More About this Item
Use for external equipment such as Aerosol Management Systems (AMS), chillers, and other supportive equipment. Inner tube and outer flange constructed of a 16 gauge 304 stainless steel. Adds 1.8" (4.6 cm) to overall width. Installation is required.
1 Year Warranty
Specifications
  • Material304 stainless steel
  • Compatible WithAll Logic® Vue biosafety enclosures
  • DescriptionLarge Tubing Pass-Through Port, 1-1/4"