Labconco Logic Vue Computer Arm
Representative image only.
Labconco Logic Vue Computer Arm
Labconco | Mfr: 3339000| Item: 3351794
Enhance the performance of your enclosure with the following accessories
₹406,000.00INR / Each
₹406,000.00INR / Each
Product Description
Includes
Articulating arm, supportive laptop platform, and mounting hardware.
Key Features
  • Eliminates need for separate computer cart
  • 1 year warranty
More About this Item
The computer arm attaches to the left side of Logic® Vue enclosures and supports monitor (up to 18 lb), keyboard, and mouse. Easily tilt the arm and move it up and down to a comfortable position. Installation is required.
1 Year Warranty
Specifications
  • Compatible WithAll Logic® Vue biosafety enclosures
  • DescriptionComputer Arm