Labconco FreeZone Three-shelf Sample Stand; 9-3/4” x 10-3/8”
Representative image only.
Labconco FreeZone Three-shelf Sample Stand; 9-3/4” x 10-3/8”
Labconco | Mfr: 7317100| Item: 0335319
₹315,000.00INR / Each
₹315,000.00INR / Each
Buy It Together
Current Item
Labconco FreeZone Three-shelf Sample Stand; 9-3/4” x 10-3/8”
Labconco FreeZone Three-shelf Sample Stand; 9-3/4” x 10-3/8”
0335319
₹315,000.00INR / Each
+ + =
Total Price 387900.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
three rubber feet.
Key Features
  • Safely accommodates bulk samples, microtiter plates, vials, and serum bottles
  • Removable shelf design offers flexible height spacing and quick loading
  • Compatible with most freeze drying chambers
  • 1 year warranty
More About this Item
Adapt, expand capacity, or just keep your freeze dryer working at its best with these parts and accessories.
1 Year Warranty
Specifications
  • Width (InchShort)10 53/64
  • Height (InchShort)9 3/4
  • DescriptionThree-shelf Sample Stand, 9 3/4” x 10 3/8”