Labconco FreeZone Slant Freeze Flask Holder, Adjustable, Powder-coated Steel
Representative image only.
Labconco FreeZone Slant Freeze Flask Holder, Adjustable, Powder-coated Steel
Labconco | Mfr: 7369700| Item: 0335325
₹106,000.00INR / Each
₹106,000.00INR / Each
Buy It Together
Current Item
Labconco FreeZone Slant Freeze Flask Holder, Adjustable, Powder-coated Steel
Labconco FreeZone Slant Freeze Flask Holder, Adjustable, Powder-coated Steel
0335325
₹106,000.00INR / Each
+ + =
Total Price 178900.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Adjustable angle—5° to 35°—offers greater pre-freezing surface area
  • Accommodates flasks of any size
  • 1 year warranty
More About this Item
Adapt, expand capacity, or just keep your freeze dryer working at its best with these parts and accessories.
1 Year Warranty
Specifications
  • Length (")10 21/64
  • Width (InchShort)4 1/2
  • Height (InchShort)6 5/8
  • DescriptionSlant Freeze Flask Holder, Adjustable, Powder-coated Steel