Labconco FreeZone Microwell Plate Holder, 5-3/4" x 4"
Representative image only.
Labconco FreeZone Microwell Plate Holder, 5-3/4" x 4"
Labconco | Mfr: 7369600| Item: 0335314
Provides excellent heat transfer to the microplate ensuring temperature stability
₹75,300.00INR / Each
₹75,300.00INR / Each
Product Description
Key Features
  • Securely holds a sinlge microplate tightly in place
  • Eliminates air gap on bottom for excellent heat transfer
  • Compatible with FreeZone® bulk tray dryers, product shelves, stoppering tray dryers, and tray dryers
  • 1 year warranty
More About this Item
Solid aluminum base plate holder provides temperature stability to small samples that are susceptible to meltback. Order microplate separately.
1 Year Warranty
Specifications
  • Length (")4
  • Width (InchShort)5 3/4
  • Height (InchShort)53/64
  • DescriptionMicrowell Plate Holder, Aluminum