Labconco FreeZone Freeze Drying Manifold, 8 Ports; FreeZone FD
Representative image only.
Labconco FreeZone Freeze Drying Manifold, 8 Ports; FreeZone FD
Labconco | Mfr: 7201800| Item: 0335312
Manifolds are ideal for freeze-drying contents of serum vials, ampules, or small flasks
₹686,000.00INR / Each
₹686,000.00INR / Each
Buy It Together
Current Item
Labconco FreeZone Freeze Drying Manifold, 8 Ports; FreeZone FD
Labconco FreeZone Freeze Drying Manifold, 8 Ports; FreeZone FD
0335312
₹686,000.00INR / Each
+ =
Total Price 686000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
one latching pin.
Key Features
  • Easily mounts atop FreeZone Freeze Dry Systems
  • Accommodates ½” and ¾” adapters for quick connection of flasks
  • Compatible with 1 L, 2.5 L, 4.5 L, 6 L, 8 L, 12 L, and 18 L FreeZone Freeze Dryers
  • 1 year warranty
More About this Item
Stainless steel manifolds are ideal for freeze-drying contents of serum vials, ampules, or small flasks.
1 Year Warranty
Specifications
  • DescriptionFreeze Drying Manifold, 8 Ports; FreeZone FD