Labconco FilterMate™ Organic Carbon Filter for FilterMate™ Portable Exhausters
Representative image only.
Labconco FilterMate™ Organic Carbon Filter for FilterMate™ Portable Exhausters
Labconco | Mfr: 3923400| Item: 0910151
Filter removes contaminants before returning air to room or exhausting to outside
₹85,600.00INR / Each
₹85,600.00INR / Each
Product Description
Key Features
  • Compatible with Labconco® FilterMate™ portable exhausters
  • Type AC
  • Contains 12 lb of granular ativated carbon media
More About this Item
Optimize performance of your Labconco FilterMate™ portable exhausters with appropriate filters.
Activated carbon media is suitable for organic solvents, including acetone, ethyl acetate, toluene, and xylene.
Specifications
  • Filter TypeOrganic carbon
  • DescriptionOrganic Carbon Filter for FilterMate™ Portable Exhausters