Labconco 7913900 Borosilicate Glass Tube with End Point Volume 1.5 mL

Representative image only.

Labconco 7913900 Borosilicate Glass Tube with End Point Volume 1.5 mL

Labconco | Mfr: 7913900| Item: 2685318

Offers better fit and heat transfer for faster evaporation rates

₹32,300.00INR / Each

₹32,300.00INR / Each
Product Description

Key Features

  • Designed for use with Cool-Zone timed end point determinations.
  • 1 year warranty

More About this Item

Borosilicate glass tubes have a 450-mL working volume when vortexing. Tubes with stems are used for timed end point determinations.

For use with RapidVap N2 Evaporation Systems only.

1 Year Warranty
Specifications
  • Tube Volume (mL)1.5
  • DescriptionBorosilicate Glass Tube with End Point Volume 1.5 mL
  • Warranty1 year
Reviews
Q&A