Labconco 7913808 Borosilicate Glass Tube with End Point Volume 2.0 mL; 8/PK
Representative image only.
Labconco 7913808 Borosilicate Glass Tube with End Point Volume 2.0 mL; 8/PK
Labconco | Mfr: 7913808| Item: 2685322
Offers better fit and heat transfer for faster evaporation rates
₹239,000.00INR / Pkg of 1
₹239,000.00INR / Pkg of 1
Product Description
Key Features
  • Designed for use with Cool-Zone timed end point determinations.
  • 1 year warranty
More About this Item
Borosilicate glass tubes have a 450-mL working volume when vortexing. Tubes with stems are used for timed end point determinations.
For use with RapidVap N2 Evaporation Systems only.
1 Year Warranty
Specifications
  • Capacity2.0 mL
  • Description2.0 mL endpoint stemmed vessel, 8 pk