Labconco 7553000 6-Place Microtiter Plate Rotor
Representative image only.
Labconco 7553000 6-Place Microtiter Plate Rotor
Labconco | Mfr: 7553000| Item: 1304059
Enhance the performance of your Labconco CentriVap® Vacuum Concentrator with these accessories
₹706,000.00INR / Each
₹706,000.00INR / Each
Product Description
Key Features
  • Choose replacement parts or optional accessories based on your application needs
  • Constructed of anodized aluminum
  • Six place microtiter plate makes it easy to interchange rotors
More About this Item
Choose from a wide range of rotors and other accessories suitable for use with your Labconco CentriVap® Complete™ Vacuum Concentrator.
Specifications
  • Capacity (Gal)6-standard 96 standard or 3 deep well plates
  • MaterialAnodized aluminum, acetal hub
  • Compatible WithCentriVap® Centrifugal Vacuum Concentrator systems
  • Description6-Place Microtiter Plate Rotor