Labconco 7462900 Aluminum DNA Rotor
Representative image only.
Labconco 7462900 Aluminum DNA Rotor
Labconco | Mfr: 7462900| Item: 1304091
Enhance the performance of your Labconco CentriVap® Vacuum Concentrator with these accessories
₹120,000.00INR / Each
₹120,000.00INR / Each
Product Description
Key Features
  • Choose replacement parts or optional accessories based on your application needs
  • Accommodates up to 72 0.5 mL tubes or 60 1.5 to 2 mL tubes
  • Constructed of anodized aluminum
More About this Item
Choose from a wide range of rotors and other accessories suitable for use with your Labconco CentriVap® Complete™ Vacuum Concentrator.
Specifications
  • Capacity (Gal)72-0.5 mL or 60-1.5 to 2.0 mL tube
  • MaterialAnodized aluminum, acetal hub
  • Compatible WithCentriVap® Centrifugal Vacuum Concentrator systems
  • DescriptionAluminum DNA Rotor