Labconco 5225106 PRECISE Glove Box Replacement ULPA Filter
Representative image only.
Labconco 5225106 PRECISE Glove Box Replacement ULPA Filter
Labconco | Mfr: 5225106| Item: 3476564
Personalize your glove box with these specially designed accessories
₹167,000.00INR / Each
₹167,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
This ULPA filter provides 99.999% efficiency on particles 0.3µm. Use in place of HEPA filter.
1 Year Warranty
Specifications
  • DescriptionPRECISE Glove Box Replacement ULPA Filter