Labconco 5005602 Glove Box Gloves, Hypalon, Size 9.75, 30"L; 1 Pair
Representative image only.
Labconco 5005602 Glove Box Gloves, Hypalon, Size 9.75, 30"L; 1 Pair
Labconco | Mfr: 5005602| Item: 3476063
Excellent glove box glove replacement
₹139,000.00INR / Pair of 1
₹139,000.00INR / Pair of 1
Product Description
Includes
O-rings.
Key Features
  • 1 year warranty
More About this Item
Order your replacement Hypalon, butyl, or neoprene gloves.
Butyl gloves are highly impermeable to water vapor, gases, and toxic chemicals.
Hypalon gloves are resistant to abrasions, ozone, and oxidizing chemicals.
Neoprene gloves are lightweight and embossed with firmhold finish.
All gloves are one-piece seamless, 0.015 gauge, come with O-rings, and fit 8 to 8 3/4" openings.
1 Year Warranty
Specifications
  • Length (")30
  • MaterialHypalon®
  • Thickness (mil)15
  • Size9.75
  • Length (in)30
  • SterileNo
  • DescriptionGlove Box Gloves, Hypalon, size 9.75, 30"L; 1 Pair