Labconco 3981401 Trace Odor Formaldehyde Carbon Filter for 5-ft Xpert® Filtered Systems (2 Required)
Representative image only.
Labconco 3981401 Trace Odor Formaldehyde Carbon Filter for 5-ft Xpert® Filtered Systems (2 Required)
Labconco | Mfr: 3981401| Item: 0910965
Trap nonvolatile nuisance odors and vapors from samples
₹116,000.00INR / Each
₹116,000.00INR / Each
Product Description
Key Features
  • Carbon filters traps odors and minute quantities of chemicals
  • Compatible with 5-ft Xpert filtered systems—requires 2 filters
  • 9.5 lb impregnated carbon
More About this Item
Easily install atop the XPert filtered systems to trap odors and small quantities of chemical vapors.
Conformite EuropeenneETL ListedISO9001:2000 Certified Supplier
Specifications
  • Compatible With5-ft Xpert® filtered systems (2 required)
  • TypeFormaldehyde carbon filter
  • DescriptionTrace Odor Formaldehyde Carbon Filter for 5-ft Xpert® Filtered Systems (2 Required)