Labconco 3937402 Trace Odor Ammonia & Amines Carbon Filter for 4-ft Xpert® Filtered Systems
Representative image only.
Labconco 3937402 Trace Odor Ammonia & Amines Carbon Filter for 4-ft Xpert® Filtered Systems
Labconco | Mfr: 3937402| Item: 0910974
Trap nonvolatile nuisance odors and vapors from samples
₹216,000.00INR / Each
₹216,000.00INR / Each
Product Description
Key Features
  • Carbon filters traps odors and minute quantities of chemicals
  • Compatible with 4-ft Xpert filtered systems
  • 12.9 lb activated carbon
More About this Item
Easily install atop the XPert filtered systems to trap odors and small quantities of chemical vapors.
Conformite EuropeenneETL ListedISO9001:2000 Certified Supplier
Specifications
  • Compatible With4-ft Xpert® filtered systems
  • TypeAmmonia & amines carbon filter
  • DescriptionTrace Odor Ammonia & Amines Carbon Filter for 4-ft Xpert® Filtered Systems