Labconco 3730600 Telescoping base stand for Biosafety Cabinets, 6-ft
Representative image only.
Labconco 3730600 Telescoping base stand for Biosafety Cabinets, 6-ft
Labconco | Mfr: 3730600| Item: 0909572
Optimize usage of your Labconco Biosafety Cabinets
₹314,000.00INR / Each
₹314,000.00INR / Each
Product Description
Key Features
  • Build a complete Biosafety Cabinet system with these accessories
  • 1 year warranty
More About this Item
Choose from a wide range of stands and accessories to ensure top performance of your Labconco Biosafety Cabinets.
Epoxy-coated steel basze stands are adjustable in 1" increments to provide a working height from 30" to 36" - and feature leveling feet.
1 Year Warranty
Specifications
  • DescriptionTelescoping base stand for Biosafety Cabinets, 6-ft