Labconco 32-Pin Rack Insert for SteamScrubber
Representative image only.
Labconco 32-Pin Rack Insert for SteamScrubber
Labconco | Mfr: 4401801| Item: 9896268
₹64,100.00INR / Each
₹64,100.00INR / Each
Buy It Together
Current Item
Labconco 32-Pin Rack Insert for SteamScrubber
Labconco 32-Pin Rack Insert for SteamScrubber
9896268
₹64,100.00INR / Each
+ +
Labconco 4401901 Accessory Retainer top for pin inserts
9896270
₹24,500.00INR / Each
=
Total Price 568600.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Configure your washer with optional inserts
  • Pins support flasks, beakers, bottles, and other dishes in an upright position
  • 1 year warranty
More About this Item
Customize your washer for your labware needs. The SteamScrubber labware washer has an upper and lower rack; however, the rack is undivided. Use these inserts to create areas for smaller labware items, especially those with narrow openings that need to be upright for thorough cleaning.
ETL Listed1 Year Warranty
Specifications
  • Description32-pin rack insert for SteamScrubber