Labconco 10-Pin Rack Insert for SteamScrubber
Representative image only.
Labconco 10-Pin Rack Insert for SteamScrubber
Labconco | Mfr: 4401501| Item: 9896266
₹32,700.00INR / Each
₹32,700.00INR / Each
Buy It Together
Current Item
Labconco 10-Pin Rack Insert for SteamScrubber
Labconco 10-Pin Rack Insert for SteamScrubber
9896266
₹32,700.00INR / Each
+ +
Labconco 4401901 Accessory Retainer top for pin inserts
9896270
₹24,500.00INR / Each
=
Total Price 537200.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Configure your washer with optional inserts
  • Pins support flasks, beakers, bottles, and other dishes in an upright position
  • 1 year warranty
More About this Item
Customize your washer for your labware needs. The SteamScrubber labware washer has an upper and lower rack; however, the rack is undivided. Use these inserts to create areas for smaller labware items, especially those with narrow openings that need to be upright for thorough cleaning.
1 Year Warranty
Specifications
  • Description10-pin rack insert for SteamScrubber