Lab-Crest 110-957-0001 Needle Valve Adapter with Neoprene O-Ring and Rubber Washer, 1/8” NPT
Representative image only.
Lab-Crest 110-957-0001 Needle Valve Adapter with Neoprene O-Ring and Rubber Washer, 1/8” NPT
Lab-Crest | Mfr: 110-957-0001| Item: 6340080
Optimize performance of your Lab-Crest pressure reaction vessels
₹38,200.00INR / Each
₹38,200.00INR / Each
Product Description
Key Features
  • O-rings ensure a tight seal
  • Comes with neoprene O-ring and rubber washer
  • Required for use with needle valve 63400-88—mates with complete coupling
More About this Item
Compatible for use with your Lab-Crest pressure reaction vessels.
Required for use with needle valve, mates with complete coupling.
Specifications
  • MaterialNeoprene, rubber, and SS
  • Compatible WithNeedle valve 63400-88
  • DescriptionNeedle Valve Adapter with Neoprene O-Ring and Rubber Washer, 1/8” NPT