Koflo 1/2-SA-3-6R-11 Sanitary Mixer, 1/2" Tri-clamp size, 6 elements
Representative image only.
Koflo 1/2-SA-3-6R-11 Sanitary Mixer, 1/2" Tri-clamp size, 6 elements
Koflo | Mfr: 1/2-SA-3-6R-11| Item: 0466828
₹207,000.00INR / Each
₹207,000.00INR / Each
Product Description
Key Features
  • Limited Lifetime Warranty
More About this Item
Blade Design Mixers are best for low-viscosity (<1,500 Cp) fluids. The faster the fluid moves, the faster (and more thorough) the mixing. Great for mixing products into water or water-like fluids.
Limited Lifetime Warranty
Specifications
  • Material316 Stainless Steel
  • No. Of Elements6
  • Pipe Id (in)0.37
  • Pipe Id (mm)9.4
  • Mixer Length (in)4
  • Mounting Type0.5" Tri-Clamp
  • DescriptionSanitary Mixer, 1/2" Tri-clamp size, 6 elements
  • WarrantyLTD Life Time