Koflo 1/2-SA-3-12RH-11 Sanitary Mixer, helical design, 1/2" Tri-clamp size, 12 elements
Representative image only.
Koflo 1/2-SA-3-12RH-11 Sanitary Mixer, helical design, 1/2" Tri-clamp size, 12 elements
Koflo | Mfr: 1/2-SA-3-12RH-11| Item: 0466876
₹413,000.00INR / Each
₹413,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Helical Design Mixers are best for high-viscosity (>1,500 Cp) fluids, and when fluids are moving more slowly through the mixer. Excellent for combining products into fluids such as yogurt, mayonnaise, and ice cream.
1 Year Warranty
Specifications
  • Material316 Stainless Steel
  • No. Of Elements12
  • Pipe Id (in)0.37
  • Pipe Id (mm)9.4
  • Mixer Length (in)10.5
  • Mounting Type0.5" Tri-Clamp
  • DescriptionSanitary Mixer, helical design, 1/2" Tri-clamp size, 12 elements