Koehler K20600 Penetrometer Precision Needle, SS with SS Ferrule, 2.5 g
Representative image only.
Koehler K20600 Penetrometer Precision Needle, SS with SS Ferrule, 2.5 g
Koehler | Mfr: K20600-00000| Item: 5986084
Compatible with manual and digital penetrometers
₹69,000.00INR / Each
₹69,000.00INR / Each
Product Description
Key Features
  • Precision machined cones and needles for ASTM and related methods
  • Penetration of bituminous materials
  • 1 year warranty
More About this Item
Use with penetrometers to determine the consistency of petroleum products.
1 Year Warranty
Specifications
  • MaterialSS with SS ferrule
  • Compatible WithManual and digital penetrometers
  • DescriptionPenetrometer Precision Needle, SS with SS Ferrule, 2.5 g