Koehler K17700 Penetrometer Precision Needle, SS, 2.5 g
Representative image only.
Koehler K17700 Penetrometer Precision Needle, SS, 2.5 g
Koehler | Mfr: K17700| Item: 5986092
Compatible with manual and digital penetrometers
₹194,000.00INR / Each
₹194,000.00INR / Each
Product Description
Key Features
  • Precision machined cones and needles for ASTM and related methods
  • Penetration of petroleum waxes
  • 1 year warranty
More About this Item
Use with penetrometers to determine the consistency of petroleum products.
1 Year Warranty
Specifications
  • MaterialSS
  • Compatible WithManual and digital penetrometers
  • DescriptionPenetrometer Precision Needle, SS, 2.5 g